Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTACATTTCTGCCATACATTTTT[A/G]CACAAAATAATGCTATTTCTGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600788 MIM: 612418 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCEB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCEB1 - transcription elongation factor B subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM70 - transmembrane protein 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040613.2 | 533 | UTR 3 | NP_001035703.1 | |||
NM_017866.5 | 533 | Missense Mutation | ACA,GCA | T,A 127 | NP_060336.3 |