Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCAAATGCATCAACCAAAGCAA[C/T]AGCATCTGAGCGAATCAGAGTGAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609751 | ||||||||||||||||||||
Literature Links: |
ACOX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACOX1 - acyl-CoA oxidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185039.1 | 2171 | Missense Mutation | ATT,GTT | I,V 566 | NP_001171968.1 | |
NM_004035.6 | 2171 | Missense Mutation | ATT,GTT | I,V 604 | NP_004026.2 | |
NM_007292.5 | 2171 | Missense Mutation | ATT,GTT | I,V 604 | NP_009223.2 | |
XM_011524868.2 | 2171 | Missense Mutation | ATT,GTT | I,V 536 | XP_011523170.1 | |
XM_011524869.2 | 2171 | Missense Mutation | ATT,GTT | I,V 468 | XP_011523171.1 |