Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAGTTCAATAAATCATCCTTCAT[A/G]TGCTTAACTGGGTGGACAGTGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612453 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105379198 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105379198 - uncharacterized LOC105379198 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MEGF10 - multiple EGF like domains 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256545.1 | Intron | NP_001243474.1 | ||||
NM_001308119.1 | Intron | NP_001295048.1 | ||||
NM_001308121.1 | Intron | NP_001295050.1 | ||||
NM_032446.2 | Intron | NP_115822.1 | ||||
XM_011543694.1 | Intron | XP_011541996.1 | ||||
XM_017009987.1 | Intron | XP_016865476.1 | ||||
XM_017009988.1 | Intron | XP_016865477.1 |