Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGCATGGAGCTGAGGATGAGGAG[G/T]CACCTTCACAGCAAGGTTTTTCTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF548 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF548 - zinc finger protein 548 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172773.1 | 458 | Missense Mutation | GCA,TCA | A,S 70 | NP_001166244.1 | |
NM_152909.3 | 458 | Missense Mutation | GCA,TCA | A,S 58 | NP_690873.2 | |
XM_017026316.1 | 458 | Missense Mutation | GCA,TCA | A,S 61 | XP_016881805.1 |
Set Membership: |
HapMap |