Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTATCAAATTCAGCTCAGTCATA[C/G]CAAAGAAGAATGGTAAGAGCTGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603402 MIM: 614309 | ||||||||||||||||||||
Literature Links: |
ACOX3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACOX3 - acyl-CoA oxidase 3, pristanoyl | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT44 - tRNA methyltransferase 44 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152544.2 | 769 | Missense Mutation | ACC,AGC | T,S 241 | NP_689757.2 | |
XM_005247950.3 | 769 | Missense Mutation | CCA,GCA | P,A 10 | XP_005248007.1 | |
XM_011513406.2 | 769 | Missense Mutation | ACC,AGC | T,S 241 | XP_011511708.1 | |
XM_011513407.2 | 769 | Missense Mutation | ACC,AGC | T,S 241 | XP_011511709.1 | |
XM_011513408.1 | 769 | Missense Mutation | ACC,AGC | T,S 12 | XP_011511710.1 | |
XM_011513409.2 | 769 | Missense Mutation | CCA,GCA | P,A 10 | XP_011511711.1 | |
XM_011513410.2 | 769 | Missense Mutation | ACC,AGC | T,S 241 | XP_011511712.1 | |
XM_017007811.1 | 769 | Missense Mutation | ACC,AGC | T,S 241 | XP_016863300.1 |