Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATATACTATATAGATCTATCTTCT[A/G]GAACATATTACTGAAAGCTAGGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608111 MIM: 602169 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FANCL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FANCL - Fanconi anemia complementation group L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114636.1 | 2721 | Intron | NP_001108108.1 | |||
NM_018062.3 | 2721 | Intron | NP_060532.2 | |||
XM_005264397.4 | 2721 | Intron | XP_005264454.1 | |||
XM_011532941.1 | 2721 | Intron | XP_011531243.1 | |||
XM_017004414.1 | 2721 | Intron | XP_016859903.1 | |||
XM_017004415.1 | 2721 | Intron | XP_016859904.1 |