Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCCATTGGAGACTTCCAAAGAC[C/G]AATAATAATTTCCCCAAATGTGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611124 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MFSD8 - major facilitator superfamily domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152778.2 | 1126 | Missense Mutation | CGT,GGT | R,G 385 | NP_689991.1 | |
XM_005262893.1 | 1126 | Missense Mutation | CGT,GGT | R,G 385 | XP_005262950.1 | |
XM_005262896.1 | 1126 | Missense Mutation | CGT,GGT | R,G 336 | XP_005262953.1 | |
XM_005262897.2 | 1126 | Missense Mutation | CGT,GGT | R,G 318 | XP_005262954.1 | |
XM_005262898.2 | 1126 | UTR 3 | XP_005262955.1 | |||
XM_011531830.1 | 1126 | Missense Mutation | CGT,GGT | R,G 347 | XP_011530132.1 | |
XM_011531831.1 | 1126 | Missense Mutation | CGT,GGT | R,G 280 | XP_011530133.1 | |
XM_011531832.1 | 1126 | UTR 3 | XP_011530134.1 | |||
XM_017007989.1 | 1126 | UTR 3 | XP_016863478.1 |