Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCTGAGGCCCAGCTCGGGAGCC[A/C]AGCAGGTGAGAGGTGAGGGAGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612524 MIM: 605700 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPP21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPP21 - ribonuclease P/MRP subunit p21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199120.1 | 1416 | Missense Mutation | CAA,CCA | Q,P 129 | NP_001186049.1 | |
NM_001199121.1 | 1416 | Missense Mutation | CAA,CCA | Q,P 121 | NP_001186050.1 | |
NM_024839.2 | 1416 | Missense Mutation | CAA,CCA | Q,P 121 | NP_079115.1 |
TRIM39 - tripartite motif containing 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM39-RPP21 - TRIM39-RPP21 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199119.1 | 1416 | Missense Mutation | CAA,CCA | Q,P 470 | NP_001186048.1 |