Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCCCAGATGTTCTGTTTCAAAC[G/T]AAGTGGATATTTCTGGCTGCCTTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613590 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTN2A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
BTN2A1 - butyrophilin subfamily 2 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197233.2 | Intron | NP_001184162.1 | ||||
NM_001197234.2 | Intron | NP_001184163.1 | ||||
NM_007049.4 | Intron | NP_008980.1 | ||||
NM_078476.3 | Intron | NP_510961.1 |
LOC285819 - uncharacterized LOC285819 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |