Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCGGGAGGTTCAGGAGAAGAGGC[A/T]CCGAGAAGCCCATGGGGAGAACAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605425 MIM: 604493 | ||||||||||||||||||||
Literature Links: |
GJB4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GJB4 - gap junction protein beta 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GJB5 - gap junction protein beta 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005268.3 | 754 | Missense Mutation | CAC,CTC | H,L 105 | NP_005259.1 | |
XM_005270751.3 | 754 | Missense Mutation | CAC,CTC | H,L 105 | XP_005270808.1 |