Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCCAGCCTCTGTCTCAGGTGGG[C/T]CAGCCTCAGCCTCTATCTCCCCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC157 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC157 - coiled-coil domain containing 157 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017437.3 | Intron | NP_001017437.2 | ||||
NM_001318334.1 | Intron | NP_001305263.1 | ||||
NM_001318335.1 | Intron | NP_001305264.1 | ||||
XM_006724278.3 | Intron | XP_006724341.1 | ||||
XM_006724279.3 | Intron | XP_006724342.1 | ||||
XM_006724280.3 | Intron | XP_006724343.1 | ||||
XM_011530263.2 | Intron | XP_011528565.1 | ||||
XM_011530266.2 | Intron | XP_011528568.1 | ||||
XM_011530267.2 | Intron | XP_011528569.1 | ||||
XM_011530268.2 | Intron | XP_011528570.1 | ||||
XM_011530269.2 | Intron | XP_011528571.1 | ||||
XM_011530270.2 | Intron | XP_011528572.1 |
KIAA1656 - KIAA1656 protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF215 - ring finger protein 215 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |