Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGGTTCGTCCTAAAAAGCTTCC[C/T]CTGATACAGAAGCGAGATACTGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607310 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM7 - ADAM metallopeptidase domain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003817.3 | 108 | Silent Mutation | CCC,CCT | P,P 36 | NP_003808.2 | |
XM_017013943.1 | 108 | Silent Mutation | CCC,CCT | P,P 36 | XP_016869432.1 |
LOC101929294 - uncharacterized LOC101929294 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |