Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTTCCATGGGCCCAGGGTAGGG[C/T]GGTGGTGGGGGCTGCACGTATCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608897 MIM: 606962 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UNC13D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UNC13D - unc-13 homolog D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WBP2 - WW domain binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012478.3 | 503 | Silent Mutation | CCA,CCG | P,P 198 | NP_036610.2 | |
XM_011524580.2 | 503 | Silent Mutation | CCA,CCG | P,P 153 | XP_011522882.1 |