Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACCTCTATGCCTGCTTGGTGGAC[A/T]TCTTCTCTGGCTGCTCCCGTTACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NT5DC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NT5DC4 - 5'-nucleotidase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011512262.2 | 2364 | Missense Mutation | ATC,TTC | I,F 231 | XP_011510564.1 | |
XM_011512265.2 | 2364 | Missense Mutation | ATC,TTC | I,F 24 | XP_011510567.1 | |
XM_017005474.1 | 2364 | Missense Mutation | ATC,TTC | I,F 231 | XP_016860963.1 | |
XM_017005475.1 | 2364 | Missense Mutation | ATC,TTC | I,F 231 | XP_016860964.1 | |
XM_017005476.1 | 2364 | Missense Mutation | ATC,TTC | I,F 167 | XP_016860965.1 | |
XM_017005477.1 | 2364 | Missense Mutation | ATC,TTC | I,F 143 | XP_016860966.1 | |
XM_017005478.1 | 2364 | Missense Mutation | ATC,TTC | I,F 124 | XP_016860967.1 |