Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCTCAGGCCCTGCTGGGCTCC[C/T]GCTCTGAGGGCTGCGACCTTCATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186820 MIM: 602602 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD7 - CD7 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SECTM1 - secreted and transmembrane 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003004.2 | 1017 | Silent Mutation | GCA,GCG | A,A 196 | NP_002995.1 | |
XM_005256392.3 | 1017 | Silent Mutation | GCA,GCG | A,A 196 | XP_005256449.1 | |
XM_011523588.2 | 1017 | Silent Mutation | GCA,GCG | A,A 196 | XP_011521890.1 |