Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTTTCCTCCCGACACGAGTGCA[C/G]TAAAGAAAAACGGTTATGAGCGTAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614484 MIM: 614724 | |||||||||||||||||||||||
Literature Links: |
ANAPC13 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ANAPC13 - anaphase promoting complex subunit 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242374.1 | 91 | UTR 5 | NP_001229303.1 | |||
NM_001242375.1 | 91 | UTR 5 | NP_001229304.1 | |||
NM_015391.3 | 91 | Intron | NP_056206.1 |