Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATAAATTAATTTCTGTAGGACTT[C/T]ATATTAACAATTTATTCATATTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614698 MIM: 602869 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COX20 - COX20, cytochrome c oxidase assembly factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001312871.1 | 13077 | Intron | NP_001299800.1 | |||
NM_001312872.1 | 13077 | Intron | NP_001299801.1 | |||
NM_001312873.1 | 13077 | Intron | NP_001299802.1 | |||
NM_001312874.1 | 13077 | Intron | NP_001299803.1 | |||
NM_198076.5 | 13077 | Intron | NP_932342.1 |
HNRNPU - heterogeneous nuclear ribonucleoprotein U | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004501.3 | 13077 | Intron | NP_004492.2 | |||
NM_031844.2 | 13077 | Intron | NP_114032.2 | |||
XM_017001115.1 | 13077 | UTR 3 | XP_016856604.1 | |||
XM_017001116.1 | 13077 | UTR 3 | XP_016856605.1 | |||
XM_017001117.1 | 13077 | UTR 3 | XP_016856606.1 |