Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCTTTGTAGTTACATTCCAGCG[C/G]GATTCTGGCTCTTCTAGACTGTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612256 MIM: 609553 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAST1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAST1 - microtubule associated serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RTBDN - retbindin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080997.2 | Intron | NP_001074466.1 | ||||
NM_001270440.1 | Intron | NP_001257369.1 | ||||
NM_001270441.1 | Intron | NP_001257370.1 | ||||
NM_001270442.1 | Intron | NP_001257371.1 | ||||
NM_001270443.1 | Intron | NP_001257372.1 | ||||
NM_001270444.1 | Intron | NP_001257373.1 | ||||
NM_001270445.1 | Intron | NP_001257374.1 | ||||
NM_031429.2 | Intron | NP_113617.1 | ||||
XM_005260089.1 | Intron | XP_005260146.1 | ||||
XM_005260090.3 | Intron | XP_005260147.1 | ||||
XM_011528343.1 | Intron | XP_011526645.1 | ||||
XM_011528344.2 | Intron | XP_011526646.1 | ||||
XM_011528345.1 | Intron | XP_011526647.1 | ||||
XM_017027343.1 | Intron | XP_016882832.1 | ||||
XM_017027344.1 | Intron | XP_016882833.1 |