Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAACTTAAATTATATAAAACCAT[A/G]ATATATAATGTTGGTAGAAGTCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606189 MIM: 604826 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRIM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRIM1 - cysteine rich transmembrane BMP regulator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FEZ2 - fasciculation and elongation protein zeta 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042548.1 | Intron | NP_001036013.1 | ||||
NM_005102.2 | Intron | NP_005093.2 | ||||
XM_006712153.2 | Intron | XP_006712216.1 |