Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCGGCGGCCACAGCTGGGGCCGG[G/T]GGCTCCGGAACGGTAAGGGCAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123834 MIM: 609514 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCND3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCND3 - cyclin D3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAF8 - TATA-box binding protein associated factor 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138572.2 | 51 | Silent Mutation | GGG,GGT | G,G 11 | NP_612639.2 | |
XM_005248849.2 | 51 | Silent Mutation | GGG,GGT | G,G 11 | XP_005248906.1 | |
XM_011514296.1 | 51 | Silent Mutation | GGG,GGT | G,G 11 | XP_011512598.1 | |
XM_017010241.1 | 51 | Silent Mutation | GGG,GGT | G,G 11 | XP_016865730.1 | |
XM_017010242.1 | 51 | Intron | XP_016865731.1 | |||
XM_017010243.1 | 51 | Intron | XP_016865732.1 | |||
XM_017010244.1 | 51 | Silent Mutation | GGG,GGT | G,G 11 | XP_016865733.1 | |
XM_017010245.1 | 51 | Silent Mutation | GGG,GGT | G,G 11 | XP_016865734.1 |