Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATACAAAGAACTGTGCTGCTTTC[C/T]TTAGACTCCACTTCCTAGGGGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616261 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PUS7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PUS7 - pseudouridylate synthase 7 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318163.1 | Intron | NP_001305092.1 | ||||
NM_001318164.1 | Intron | NP_001305093.1 | ||||
NM_019042.4 | Intron | NP_061915.2 | ||||
XM_005250462.3 | Intron | XP_005250519.1 | ||||
XM_011516336.2 | Intron | XP_011514638.1 | ||||
XM_017012367.1 | Intron | XP_016867856.1 | ||||
XM_017012368.1 | Intron | XP_016867857.1 |