Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTAGATGGGGAGACTGGGACACA[C/T]GTCTGAACGCAGATGTCCGAAGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612665 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEX101 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TEX101 - testis expressed 101 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130011.1 | Intron | NP_001123483.1 | ||||
NM_031451.4 | Intron | NP_113639.4 | ||||
XM_005259303.3 | Intron | XP_005259360.2 |