Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAATACACTGGGCAGACATAAA[C/G]AGGTAGGGGAGTCAGATGTCCTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602670 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLE2 - DNA polymerase epsilon 2, accessory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197330.1 | 1543 | Missense Mutation | CTT,GTT | L,V 430 | NP_001184259.1 | |
NM_001197331.1 | 1543 | Missense Mutation | CTT,GTT | L,V 456 | NP_001184260.1 | |
NM_002692.3 | 1543 | Missense Mutation | CTT,GTT | L,V 456 | NP_002683.2 | |
XM_011536842.2 | 1543 | Missense Mutation | CTT,GTT | L,V 430 | XP_011535144.1 | |
XM_011536843.2 | 1543 | Missense Mutation | CTT,GTT | L,V 379 | XP_011535145.1 | |
XM_011536844.1 | 1543 | Intron | XP_011535146.1 | |||
XM_017021374.1 | 1543 | Missense Mutation | CTT,GTT | L,V 456 | XP_016876863.1 | |
XM_017021375.1 | 1543 | Missense Mutation | CTT,GTT | L,V 379 | XP_016876864.1 |