Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATTCTCCTTGAGAAGCAGCTAT[A/T]ATCAACAGAACATTGACAGAACCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615256 | ||||||||||||||||||||
Literature Links: |
AMN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AMN1 - antagonist of mitotic exit network 1 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ETFBKMT - electron transfer flavoprotein beta subunit lysine methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135863.1 | 246 | UTR 5 | NP_001129335.1 | |||
NM_001135864.1 | 246 | UTR 5 | NP_001129336.1 | |||
NM_173802.3 | 246 | UTR 5 | NP_776163.1 |