Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616759 MIM: 616633 MIM: 604429 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOSIP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NOSIP - nitric oxide synthase interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR12 - proline rich 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRRG2 - proline rich and Gla domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000951.2 | 425 | Silent Mutation | TTC,TTT | F,F 31 | NP_000942.1 | |
NM_001316335.1 | 425 | Silent Mutation | TTC,TTT | F,F 8 | NP_001303264.1 | |
XM_006723286.2 | 425 | Silent Mutation | TTC,TTT | F,F 31 | XP_006723349.1 | |
XM_011527124.1 | 425 | Silent Mutation | TTC,TTT | F,F 8 | XP_011525426.1 |
Set Membership: |
HapMap |