Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGGAGAAAAAACTATGAACTAC[G/T]TTGCTTTCACCAGTGTAAACACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610211 MIM: 603055 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLIRP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLIRP - SRA stem-loop interacting RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267863.1 | Intron | NP_001254792.1 | ||||
NM_001267864.1 | Intron | NP_001254793.1 | ||||
NM_031210.5 | Intron | NP_112487.1 |
SNW1 - SNW domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318844.1 | Intron | NP_001305773.1 | ||||
NM_012245.2 | Intron | NP_036377.1 | ||||
XM_005267413.3 | Intron | XP_005267470.1 |
Set Membership: |
HapMap |