Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAATACTAATACACAGACAATG[C/T]AACATTGTATCATTTACAAATCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613542 | ||||||||||||||||||||
Literature Links: |
MTRF1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTRF1L - mitochondrial translational release factor 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114184.2 | Intron | NP_001107656.1 | ||||
NM_001301047.2 | Intron | NP_001287976.1 | ||||
NM_001301870.1 | Intron | NP_001288799.1 | ||||
NM_001301871.1 | Intron | NP_001288800.1 | ||||
NM_001301872.1 | Intron | NP_001288801.1 | ||||
NM_019041.6 | Intron | NP_061914.3 | ||||
XM_017010959.1 | Intron | XP_016866448.1 | ||||
XM_017010960.1 | Intron | XP_016866449.1 |