Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGCTTGTTTCCCGTTGCCCAGG[A/G]CTGTCGGGTCCCTGTTCAGGCCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMTK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LMTK3 - lemur tyrosine kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080434.1 | 3144 | Missense Mutation | GCC,GTC | A,V 938 | NP_001073903.1 | |
XM_011526411.2 | 3144 | Missense Mutation | GCC,GTC | A,V 935 | XP_011524713.1 | |
XM_011526412.2 | 3144 | Missense Mutation | GCC,GTC | A,V 924 | XP_011524714.1 |