Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCGCGCGGGCAGGCAGACGGGG[C/T]GCACGGCGGCCGAGCGCACCACCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616386 MIM: 609862 | ||||||||||||||||||||
Literature Links: |
KCTD17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCTD17 - potassium channel tetramerization domain containing 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMPRSS6 - transmembrane protease, serine 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289000.1 | 1795 | Missense Mutation | CAC,CGC | H,R 676 | NP_001275929.1 | |
NM_001289001.1 | 1795 | Missense Mutation | CAC,CGC | H,R 676 | NP_001275930.1 | |
NM_153609.3 | 1795 | Missense Mutation | CAC,CGC | H,R 685 | NP_705837.1 | |
XM_011529989.2 | 1795 | Missense Mutation | CAC,CGC | H,R 532 | XP_011528291.1 |