Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGTCACAGGCTTCCGTCAGCTC[A/G]GAAATCCCACCCACCTCTGTGGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126063 | ||||||||||||||||||||
Literature Links: |
DLST PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLST - dihydrolipoamide S-succinyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS6KL1 - ribosomal protein S6 kinase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031464.4 | 1500 | Silent Mutation | TCC,TCT | S,S 459 | NP_113652.2 | |
XM_011537206.2 | 1500 | Silent Mutation | TCC,TCT | S,S 475 | XP_011535508.1 | |
XM_011537207.2 | 1500 | Silent Mutation | TCC,TCT | S,S 475 | XP_011535509.1 | |
XM_011537211.2 | 1500 | Silent Mutation | TCC,TCT | S,S 428 | XP_011535513.1 | |
XM_011537213.2 | 1500 | Silent Mutation | TCC,TCT | S,S 412 | XP_011535515.1 | |
XM_017021680.1 | 1500 | Silent Mutation | TCC,TCT | S,S 475 | XP_016877169.1 | |
XM_017021681.1 | 1500 | Silent Mutation | TCC,TCT | S,S 459 | XP_016877170.1 | |
XM_017021682.1 | 1500 | Silent Mutation | TCC,TCT | S,S 459 | XP_016877171.1 | |
XM_017021683.1 | 1500 | Silent Mutation | TCC,TCT | S,S 445 | XP_016877172.1 | |
XM_017021684.1 | 1500 | Silent Mutation | TCC,TCT | S,S 475 | XP_016877173.1 | |
XM_017021685.1 | 1500 | Silent Mutation | TCC,TCT | S,S 475 | XP_016877174.1 | |
XM_017021686.1 | 1500 | Nonsense Mutation | CGA,TGA | R,* 466 | XP_016877175.1 | |
XM_017021687.1 | 1500 | Nonsense Mutation | CGA,TGA | R,* 466 | XP_016877176.1 | |
XM_017021688.1 | 1500 | Silent Mutation | TCC,TCT | S,S 273 | XP_016877177.1 | |
XM_017021689.1 | 1500 | Silent Mutation | TCC,TCT | S,S 273 | XP_016877178.1 | |
XM_017021690.1 | 1500 | Silent Mutation | TCC,TCT | S,S 257 | XP_016877179.1 | |
XM_017021691.1 | 1500 | Silent Mutation | TCC,TCT | S,S 196 | XP_016877180.1 | |
XM_017021692.1 | 1500 | Silent Mutation | TCC,TCT | S,S 243 | XP_016877181.1 |