Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTAAGTGTTCTGGCACTATCCA[C/G]GTTCAGAAGGTGTTGAGGCCATTAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610372 MIM: 605521 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC2A12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC2A12 - solute carrier family 2 member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145176.2 | 2047 | UTR 3 | NP_660159.1 | |||
XM_006715349.3 | 2047 | Intron | XP_006715412.1 | |||
XM_017010311.1 | 2047 | Intron | XP_016865800.1 |
TBPL1 - TATA-box binding protein like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |