Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCACTTTATATTTCTTCTAGAA[A/G]AGATCCAATTTGTCATTGTTTGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQCD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IQCD - IQ motif containing D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138451.2 | Intron | NP_612460.1 | ||||
XM_005253832.4 | Intron | XP_005253889.1 | ||||
XM_005253833.4 | Intron | XP_005253890.1 | ||||
XM_005253834.4 | Intron | XP_005253891.1 | ||||
XM_011537862.2 | Intron | XP_011536164.1 | ||||
XM_011537863.2 | Intron | XP_011536165.1 | ||||
XM_011537864.1 | Intron | XP_011536166.1 | ||||
XM_011537865.2 | Intron | XP_011536167.1 | ||||
XM_011537866.2 | Intron | XP_011536168.1 | ||||
XM_017018771.1 | Intron | XP_016874260.1 |
RITA1 - RBPJ interacting and tubulin associated 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |