Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606867 MIM: 611430 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GORASP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GORASP1 - golgi reassembly stacking protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC21A - tetratricopeptide repeat domain 21A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105513.2 | Intron | NP_001098983.2 | ||||
NM_145755.2 | Intron | NP_665698.2 | ||||
XM_005264921.4 | Intron | XP_005264978.1 | ||||
XM_005264922.4 | Intron | XP_005264979.1 | ||||
XM_005264923.3 | Intron | XP_005264980.1 | ||||
XM_005264924.4 | Intron | XP_005264981.1 | ||||
XM_005264925.4 | Intron | XP_005264982.1 | ||||
XM_005264926.4 | Intron | XP_005264983.1 | ||||
XM_005264927.4 | Intron | XP_005264984.1 | ||||
XM_006713011.3 | Intron | XP_006713074.1 | ||||
XM_011533447.2 | Intron | XP_011531749.1 | ||||
XM_011533449.1 | Intron | XP_011531751.1 | ||||
XM_017005843.1 | Intron | XP_016861332.1 |