Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGAAGGGGGAGTCGGGACCCTG[C/G]GGGGAGCTCCGTGGGCTGGCTACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616846 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EMC1 - ER membrane protein complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271427.1 | 84 | Intron | NP_001258356.1 | |||
NM_001271428.1 | 84 | Intron | NP_001258357.1 | |||
NM_001271429.1 | 84 | Intron | NP_001258358.1 | |||
NM_015047.2 | 84 | Intron | NP_055862.1 | |||
XM_005245787.1 | 84 | Intron | XP_005245844.1 | |||
XM_005245788.1 | 84 | Intron | XP_005245845.1 |
MRTO4 - MRT4 homolog, ribosome maturation factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016183.3 | 84 | Intron | NP_057267.2 | |||
XM_006710675.3 | 84 | UTR 5 | XP_006710738.1 |