Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCAACGGGCTCTTTGGAGAAAA[A/G]TCTTATGATTTCCTCACAGTAAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612086 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMSD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMSD - histocompatibility minor serpin domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123366.1 | 448 | Silent Mutation | AAA,AAG | K,K 68 | NP_001116838.1 | |
XM_011525930.2 | 448 | Silent Mutation | AAA,AAG | K,K 68 | XP_011524232.1 | |
XM_017025710.1 | 448 | Silent Mutation | AAA,AAG | K,K 68 | XP_016881199.1 |