Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACTCTGAATGGAGAGAAGATTCG[A/C]GCGCGGGTGCTCCCGGGCCAGGCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608934 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEIL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NEIL3 - nei like DNA glycosylase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018248.2 | 128 | Silent Mutation | CGA,CGC | R,R 15 | NP_060718.2 | |
XM_017008360.1 | 128 | Silent Mutation | CGA,CGC | R,R 15 | XP_016863849.1 |
Set Membership: |
HapMap |