Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCTACTCCCACCCCCTGGCCCT[A/G]GTCAAACTCACATTTGGGGTTTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHX37 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DHX37 - DEAH-box helicase 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032656.3 | 3503 | Intron | NP_116045.2 | |||
XM_005253590.3 | 3503 | Nonsense Mutation | CAG,TAG | Q,* 1134 | XP_005253647.1 | |
XM_011538598.2 | 3503 | Intron | XP_011536900.1 | |||
XM_011538600.2 | 3503 | Intron | XP_011536902.1 |
Set Membership: |
HapMap |