Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGACCCTGAAATTTTACAAAAA[A/T]TTTTTGGTATTTATTTTCATATGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM78B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM78B - family with sequence similarity 78 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017961.4 | Intron | NP_001017961.1 | ||||
NM_001320302.1 | Intron | NP_001307231.1 | ||||
XM_017000417.1 | Intron | XP_016855906.1 | ||||
XM_017000418.1 | Intron | XP_016855907.1 |
LOC105371586 - uncharacterized LOC105371586 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |