Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGCACCTGTTGACTCGTTTAGGC[C/T]TAAACACCCTGGGAGTTAGGCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604517 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPARGC1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPARGC1A - PPARG coactivator 1 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013261.3 | Intron | NP_037393.1 | ||||
XM_005248131.4 | Intron | XP_005248188.1 | ||||
XM_005248132.1 | Intron | XP_005248189.1 | ||||
XM_005248134.4 | Intron | XP_005248191.1 | ||||
XM_011513765.2 | Intron | XP_011512067.1 | ||||
XM_011513766.1 | Intron | XP_011512068.1 | ||||
XM_011513767.2 | Intron | XP_011512069.1 | ||||
XM_011513768.1 | Intron | XP_011512070.1 | ||||
XM_011513769.2 | Intron | XP_011512071.1 | ||||
XM_011513770.2 | Intron | XP_011512072.1 | ||||
XM_011513771.1 | Intron | XP_011512073.1 | ||||
XM_017007664.1 | Intron | XP_016863153.1 |