Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCACCCGCAGCCTTCCAGCAGG[G/A]CAGCTCCCTGGGGCTTGGTTCAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608601 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBRS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBRS - fibrosin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105079.2 | Intron | NP_001098549.2 | ||||
XM_011545916.2 | Intron | XP_011544218.1 | ||||
XM_011545917.2 | Intron | XP_011544219.1 | ||||
XM_011545919.2 | Intron | XP_011544221.1 |
PRR14 - proline rich 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |