Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGGGCCGGAACTGCCGTCGCCGG[C/T]GCGGTCGTTGTCGCATTGCTCTCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAXD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAXD - NAD(P)HX dehydratase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242881.1 | 130 | Silent Mutation | GGC,GGT | G,G 13 | NP_001229810.1 | |
NM_001242882.1 | 130 | Intron | NP_001229811.1 | |||
NM_001242883.1 | 130 | Intron | NP_001229812.1 | |||
NM_018210.3 | 130 | Silent Mutation | GGC,GGT | G,G 13 | NP_060680.2 |