Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGGGAATGATCTATAAAAGAAAA[C/T]AATGACAATATATTAATTGACAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615256 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMN1 - antagonist of mitotic exit network 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113402.1 | Intron | NP_001106873.1 | ||||
NM_001278411.1 | Intron | NP_001265340.1 | ||||
NM_001278412.1 | Intron | NP_001265341.1 | ||||
XM_006719054.3 | Intron | XP_006719117.1 | ||||
XM_011520604.2 | Intron | XP_011518906.1 | ||||
XM_017018963.1 | Intron | XP_016874452.1 | ||||
XM_017018964.1 | Intron | XP_016874453.1 | ||||
XM_017018965.1 | Intron | XP_016874454.1 |
ETFBKMT - electron transfer flavoprotein beta subunit lysine methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135863.1 | Intron | NP_001129335.1 | ||||
NM_001135864.1 | Intron | NP_001129336.1 | ||||
NM_173802.3 | Intron | NP_776163.1 |