Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACACTCACTGCTAGAATTCAGAGT[G/T]GTGTCTTGGTAAGCCTTGTAAATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611883 MIM: 607960 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCCIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCCIP - BRCA2 and CDKN1A interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016567.3 | 2184 | Intron | NP_057651.1 | |||
NM_078468.2 | 2184 | Intron | NP_510868.1 | |||
NM_078469.2 | 2184 | Intron | NP_510869.1 |
DHX32 - DEAH-box helicase 32 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018180.2 | 2184 | Silent Mutation | ACA,ACC | T,T 557 | NP_060650.2 | |
XM_017016404.1 | 2184 | Silent Mutation | ACA,ACC | T,T 557 | XP_016871893.1 | |
XM_017016405.1 | 2184 | Silent Mutation | ACA,ACC | T,T 557 | XP_016871894.1 |