Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTTTGGAAAAAATACAGAGTTC[A/G]TATTTTTTGGGGTGATATGAGGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608684 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NIN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NIN - ninein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016350.4 | 6410 | Intron | NP_057434.4 | |||
NM_020921.3 | 6410 | Intron | NP_065972.3 | |||
NM_182944.2 | 6410 | Intron | NP_891989.2 | |||
NM_182946.1 | 6410 | Intron | NP_891991.1 | |||
XM_011536817.1 | 6410 | Intron | XP_011535119.1 | |||
XM_011536819.2 | 6410 | Intron | XP_011535121.1 | |||
XM_011536822.1 | 6410 | Intron | XP_011535124.1 | |||
XM_011536823.2 | 6410 | UTR 3 | XP_011535125.1 |