Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACACCAACACCTTCTTTCTGTGCC[A/G]GACACTGCGCTCCCTAGGGGTCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 116900 MIM: 610595 MIM: 607377 | ||||||||||||||||||||
Literature Links: |
CKS1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CKS1B - CDC28 protein kinase regulatory subunit 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FLAD1 - flavin adenine dinucleotide synthetase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184891.1 | 367 | Missense Mutation | CAG,CGG | Q,R 40 | NP_001171820.1 | |
NM_001184892.1 | 367 | Missense Mutation | CAG,CGG | Q,R 38 | NP_001171821.1 | |
NM_025207.4 | 367 | Missense Mutation | CAG,CGG | Q,R 137 | NP_079483.3 | |
NM_201398.2 | 367 | Missense Mutation | CAG,CGG | Q,R 40 | NP_958800.1 |
LENEP - lens epithelial protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |