Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAATAGCTGGGGCTCTCAGTGC[A/C]CATAGGTATTTCCGAGATAATCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTR6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTR6 - ARP6 actin-related protein 6 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022496.4 | 1161 | Silent Mutation | GCA,GCC | A,A 132 | NP_071941.1 | |
XM_017019821.1 | 1161 | Silent Mutation | GCA,GCC | A,A 24 | XP_016875310.1 |
DEPDC4 - DEP domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319310.1 | 1161 | Intron | NP_001306239.1 | |||
NM_001319311.1 | 1161 | Intron | NP_001306240.1 | |||
NM_152317.3 | 1161 | Intron | NP_689530.1 | |||
XM_011537876.2 | 1161 | Intron | XP_011536178.1 | |||
XM_017018779.1 | 1161 | Intron | XP_016874268.1 | |||
XM_017018780.1 | 1161 | Intron | XP_016874269.1 | |||
XM_017018781.1 | 1161 | Intron | XP_016874270.1 | |||
XM_017018782.1 | 1161 | Intron | XP_016874271.1 | |||
XM_017018783.1 | 1161 | Intron | XP_016874272.1 | |||
XM_017018784.1 | 1161 | Intron | XP_016874273.1 | |||
XM_017018785.1 | 1161 | Intron | XP_016874274.1 |