Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGGAGGAGGAGGACGTTCTGGC[A/G]GGGGTCTCAGCAGAGGACAAGAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601768 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPND PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MPND - MPN domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159846.2 | 664 | Silent Mutation | GCA,GCG | A,A 199 | NP_001153318.1 | |
NM_001300862.1 | 664 | Silent Mutation | GCA,GCG | A,A 199 | NP_001287791.1 | |
NM_032868.5 | 664 | Silent Mutation | GCA,GCG | A,A 199 | NP_116257.2 | |
XM_006722926.2 | 664 | Silent Mutation | GCA,GCG | A,A 199 | XP_006722989.1 |
SH3GL1 - SH3 domain containing GRB2 like 1, endophilin A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |