Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGAATGAATGACTCCTTCATGA[A/T]GACCCCCTGCACTGTGCCCTGGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600608 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
P4HA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
P4HA2 - prolyl 4-hydroxylase subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017973.1 | Intron | NP_001017973.1 | ||||
NM_001017974.1 | Intron | NP_001017974.1 | ||||
NM_001142598.1 | Intron | NP_001136070.1 | ||||
NM_001142599.1 | Intron | NP_001136071.1 | ||||
NM_004199.2 | Intron | NP_004190.1 | ||||
XM_005272116.4 | Intron | XP_005272173.1 | ||||
XM_005272117.4 | Intron | XP_005272174.1 | ||||
XM_005272118.4 | Intron | XP_005272175.1 | ||||
XM_005272119.4 | Intron | XP_005272176.1 | ||||
XM_005272120.4 | Intron | XP_005272177.1 | ||||
XM_006714728.3 | Intron | XP_006714791.1 | ||||
XM_006714729.3 | Intron | XP_006714792.1 | ||||
XM_006714730.3 | Intron | XP_006714793.1 | ||||
XM_011543705.2 | Intron | XP_011542007.1 | ||||
XM_017010011.1 | Intron | XP_016865500.1 | ||||
XM_017010012.1 | Intron | XP_016865501.1 |
P4HA2-AS1 - P4HA2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |