Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCACCACCACCGTCCGCCCCCC[C/T]TGGAAATAAGACAACTGTCCAGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
YLPM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
YLPM1 - YLP motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019589.2 | 929 | Missense Mutation | CCT,CTT | P,L 254 | NP_062535.2 | |
XM_005267860.3 | 929 | Missense Mutation | CCT,CTT | P,L 254 | XP_005267917.1 | |
XM_011536966.2 | 929 | Intron | XP_011535268.1 | |||
XM_011536967.1 | 929 | Missense Mutation | CCT,CTT | P,L 254 | XP_011535269.1 |