Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCAGAGCCCCCAGCCAGGCCAG[C/T]CCCACGTGTGCTGCGGGGTGGGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606669 MIM: 606684 | ||||||||||||||||||||
Literature Links: |
FXYD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FXYD5 - FXYD domain containing ion transport regulator 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164605.1 | Intron | NP_001158077.1 | ||||
NM_001320912.1 | Intron | NP_001307841.1 | ||||
NM_001320913.1 | Intron | NP_001307842.1 | ||||
NM_014164.5 | Intron | NP_054883.3 | ||||
NM_144779.2 | Intron | NP_659003.1 |
FXYD7 - FXYD domain containing ion transport regulator 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |